Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2,3-difluorobenzaldehyde, 98%

Catalog No. AAH64044MD
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 644985-24-0 | C7H3BrF2O | 221.001 g/mol

4-Bromo-2,3-difluorobenzaldehyde is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2,3-difluorobenzaldehyde is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 644985-24-0
Molecular Formula C7H3BrF2O
Molecular Weight (g/mol) 221.001
MDL Number MFCD09258953
InChI Key IMOLPSNRLZLWQR-UHFFFAOYSA-N
Synonym 2,3-difluoro-4-bromobenzaldehyde, benzaldehyde, 4-bromo-2,3-difluoro, 1-bromo-2,3-difluoro-4-formylbenzene, pubchem10068, 4-bromo-2,3-difluoro-benzaldehyde, 4-bromanyl-2,3-bis fluoranyl benzaldehyde, 4-bromo-2, 3-difluorobenzaldehyde
PubChem CID 26986390
IUPAC Name 4-bromo-2,3-difluorobenzaldehyde
SMILES C1=CC(=C(C(=C1C=O)F)F)Br

Specifications

Melting Point 66°C to 67°C
Color White
Quantity 250 mg
Beilstein 9551272
Solubility Information Slightly soluble in water.
Formula Weight 221
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Bromo-2,3-difluorobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.