Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxypropyl methacrylate, 97+%, mixture of isomers, stabilized

Catalog No. AC412150010
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle

CAS: 27813-02-1 | C7H12O3 | 144.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27813-02-1
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00004536
InChI Key ZMARGGQEAJXRFP-UHFFFAOYNA-N
Synonym 2-hydroxypropyl methacrylate, 2-hydroxypropylmethacrylate, hpma, beta-hydroxypropyl methacrylate, acryester hp, 2-hydroxypropyl 2-methylacrylate, poly 2-hydroxypropyl methacrylate, 2-hydroxypropyl 2-methyl-2-propenoate, 2-propenoic acid, 2-methyl-, 2-hydroxypropyl ester, 2-hpma
PubChem CID 13539
ChEBI CHEBI:53440
IUPAC Name 2-hydroxypropyl 2-methylprop-2-enoate
SMILES CC(CO)OC(=O)C(C)=C

Specifications

CAS Min % 97.0
CAS Max % 100.0
Density 1.0300g/mL
Boiling Point 240.0°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (Mixture of isomers) (GC)
Packaging Glass bottle
Refractive Index 1.4470 to 1.4490
Linear Formula CH2=C(CH3)CO2CH2CH(OH)CH3
Quantity 1 kg
Specific Gravity 1.03
Solubility Information Solubility in water: 130g/L (25°C). Other solubilities: miscible in almost all organic solvents
Stabilizer 150 to 300ppm MEHQ
Viscosity 9.9 mm2/s (20°C)
Formula Weight 144.17
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material Hydroxypropyl methacrylate, 96.5%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
EINECSNumber 248-666-3
RTECSNumber OZ4750000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.