Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2,6-dimethylaniline, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
250g
Packaging:
Glass bottle
24596-19-8
C8H10BrN
200.08
MFCD00007826
QGLAYJCJLHNIGJ-UHFFFAOYSA-N
4-bromo-2,6-xylidine,benzenamine, 4-bromo-2,6-dimethyl,4-bromo-2,6-dimethyl-phenylamine,2-amino-5-bromo-m-xylene,2,6-dimethyl-4-bromoaniline,4-bromo-2,6-dimethylbenzenamine,4-bromo-2,6-dimethylphenylamine,4-bromo-2,6-dimethyl-aniline,4-bromo-2,6-dimethylaniline,4-bromo-2,6-dimethyl-benzenamine
90549
4-bromo-2,6-dimethylaniline
CC1=CC(=CC(=C1N)C)Br
24596-19-8
C8H10BrN
200.08
MFCD00007826
QGLAYJCJLHNIGJ-UHFFFAOYSA-N
4-bromo-2,6-xylidine,benzenamine, 4-bromo-2,6-dimethyl,4-bromo-2,6-dimethyl-phenylamine,2-amino-5-bromo-m-xylene,2,6-dimethyl-4-bromoaniline,4-bromo-2,6-dimethylbenzenamine,4-bromo-2,6-dimethylphenylamine,4-bromo-2,6-dimethyl-aniline,4-bromo-2,6-dimethylaniline,4-bromo-2,6-dimethyl-benzenamine
90549
4-bromo-2,6-dimethylaniline
CC1=CC(=CC(=C1N)C)Br
Color Amber to Yellow
Packaging Glass bottle
Flash Point >110°C
Quantity 5g
Melting Point 48.0°C to 52.0°C
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula BrC6H2(CH3)2NH2
Beilstein 12,111
Solubility Information Solubility in water: insoluble
Formula Weight 200.08
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 4-Bromo-2, 6-dimethylaniline, 98%
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a P

GHS Signal Word: Warning

EINECSNumber : 246-337-9