Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-chlorophenol, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
3964-56-5
C6H4BrClO
207.45
MFCD00002166
VIBJPUXLAKVICD-UHFFFAOYSA-N
2-chloro-4-bromophenol,phenol, 4-bromo-2-chloro,phenol, 4-bromochloro,unii-994sj34d8p,4-bromo-2-chloro-phenol,attercop-chm at106179,4-bromo-2-chlorophenol 25gr,4-bromo-chlorophenol,pubchem3596,acmc-209j6p
19859
CHEBI:38852
4-bromo-2-chlorophenol
C1=CC(=C(C=C1Br)Cl)O
3964-56-5
C6H4BrClO
207.45
MFCD00002166
VIBJPUXLAKVICD-UHFFFAOYSA-N
2-chloro-4-bromophenol,phenol, 4-bromo-2-chloro,phenol, 4-bromochloro,unii-994sj34d8p,4-bromo-2-chloro-phenol,attercop-chm at106179,4-bromo-2-chlorophenol 25gr,4-bromo-chlorophenol,pubchem3596,acmc-209j6p
19859
CHEBI:38852
4-bromo-2-chlorophenol
C1=CC(=C(C=C1Br)Cl)O
Infrared Spectrum Authentic
Boiling Point 232°C to 235°C
Flash Point >112°C
Melting Point 47°C to 51°C
Color White
Quantity 5g
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula BrC6H3(Cl)OH
Beilstein 06, II, 187
Formula Weight 207.45
Physical Form Low Melting Mass or Crystalline Needles, Powder and Chunks
Percent Purity 99%
Chemical Name or Material 4-Bromo-2-chlorophenol, 99%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Toxic if swallowed.
Causes skin irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 223-572-5