Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-chlorophenol, 99%

Catalog No. AC173530250
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 3964-56-5 | C6H4BrClO | 207.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3964-56-5
Molecular Formula C6H4BrClO
Molecular Weight (g/mol) 207.45
MDL Number MFCD00002166
InChI Key VIBJPUXLAKVICD-UHFFFAOYSA-N
Synonym 2-chloro-4-bromophenol, phenol, 4-bromo-2-chloro, phenol, 4-bromochloro, unii-994sj34d8p, 4-bromo-2-chloro-phenol, attercop-chm at106179, 4-bromo-2-chlorophenol 25gr, 4-bromo-chlorophenol, pubchem3596, acmc-209j6p
PubChem CID 19859
ChEBI CHEBI:38852
IUPAC Name 4-bromo-2-chlorophenol
SMILES OC1=CC=C(Br)C=C1Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 47°C to 51°C
Color White
Boiling Point 232°C to 235°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H3(Cl)OH
Quantity 25 g
Beilstein 06, II, 187
Formula Weight 207.45
Percent Purity 99%
Physical Form Low Melting Mass or Crystalline Needles, Powder and Chunks
Chemical Name or Material 4-Bromo-2-chlorophenol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Toxic if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 223-572-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.