Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-fluoro-5-(trifluoromethyl)aniline, 97%

Catalog No. aah3172709
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3172709 10 g
AAH3172703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3172709 Supplier Thermo Scientific Chemicals Supplier No. H3172709
Only null left
Add to Cart

CAS: 104460-70-0 | C7H4BrF4N | 258.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Bromo-2-fluoro-5-(trifluoromethyl)aniline has antimicrobial effect.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104460-70-0
Molecular Formula C7H4BrF4N
Molecular Weight (g/mol) 258.01
MDL Number MFCD04973757
InChI Key UYVDMCXPDGRLEC-UHFFFAOYSA-N
Synonym 4-bromo-2-fluoro-5-trifluoromethyl aniline, benzenamine, 4-bromo-2-fluoro-5-trifluoromethyl, benzenamine,4-bromo-2-fluoro-5-trifluoromethyl, acmc-209ygd, 4-bromo-2-fluoro-5-trifluoromethyl phenylamine, 4-bromanyl-2-fluoranyl-5-trifluoromethyl aniline
PubChem CID 7018046
IUPAC Name 4-bromo-2-fluoro-5-(trifluoromethyl)aniline
SMILES NC1=C(F)C=C(Br)C(=C1)C(F)(F)F

Specifications

Melting Point 30°C to 41°C
Quantity 10 g
UN Number UN2811
Formula Weight 258.01
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-fluoro-5-(trifluoromethyl)aniline
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.