Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-fluoroanisole 98.0+%, TCI America™
SDP

Catalog No. B185525G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 2357-52-0
Molecular Formula C7H6BrFO
Molecular Weight (g/mol) 205.026
MDL Number MFCD00011710
InChI Key DWNXGZBXFDNKOR-UHFFFAOYSA-N
Synonym 4-bromo-2-fluoroanisole, 2-fluoro-4-bromoanisole, 1-bromo-3-fluoro-4-methoxybenzene, benzene, 4-bromo-2-fluoro-1-methoxy, 2-fluoro-4-bromo anisol, 2-fluoro-4-bromo anisole, 4-bromo-2-fluoro-1-methoxy-benzene, 4-bromo-2-fluorophenylmethyl ether, 4-brom-2-fluor-1-methoxybenzol, pubchem1971
PubChem CID 75378
IUPAC Name 4-bromo-2-fluoro-1-methoxybenzene
SMILES COC1=C(C=C(C=C1)Br)F

Specifications

Color Yellow
Boiling Point 208°C
Quantity 25 g
Formula Weight 205.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-2-fluoroanisole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.