Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-nitroaniline, 98%

Catalog No. p-7028252
Click to view available options
Quantity:
25 g
100 g

CAS: 875-51-4 | C6H5BrN2O2 | 217.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 875-51-4
Molecular Formula C6H5BrN2O2
Molecular Weight (g/mol) 217.02
MDL Number MFCD00041312
InChI Key ZCWBZRBJSPWUPG-UHFFFAOYSA-N
Synonym benzenamine, 4-bromo-2-nitro, 2-nitro-4-bromoaniline, 4-bromo-o-nitroaniline, p-bromo-o-nitroaniline, 4-bromo-2-nitro-phenylamine, 4-bromo-2-nitrobenzeneamine, aniline, 4-bromo-2-nitro, 4-bromo-2-nitrobenzenamine, 1-amino-4-bromo-2-nitrobenzene, 4-bromo-2-nitro-aniline
PubChem CID 70132
IUPAC Name 4-bromo-2-nitroaniline
SMILES NC1=CC=C(Br)C=C1[N+]([O-])=O

Specifications

Melting Point 111°C to 114°C
Quantity 25 g
Formula Weight 217.02
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-nitroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
RTECSNumber BW9370000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.