Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Bromo-2-nitrotoluene, 98%, Thermo Scientific™
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 43.0°C to 46.0°C |
Color | Orange to Yellow |
Boiling Point | 130.0°C (12.0 mmHg) |
Flash Point | >110°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Quantity | 5 g |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Causes serious eye irritation. Harmful if inhaled. May cause respiratory irritation. Harmful in contact with skin. Causes skin irritation. Harmful if swallowed. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for |
EINECSNumber | 262-536-3 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction