Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-thiophenecarbonyl chloride, 97%, Thermo Scientific™

Catalog No. CC28502DA
Click to view available options
:
Amber Glass Bottle; 1g

Specifications

CAS 58777-65-4
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C5H2BrClOS
Quantity 1g
MDL Number MFCD04115391
Synonym 4-bromo-2-thiophenecarbonyl chloride, 3-bromo-5-thiophencarbonyl chloride, 2-thiophenecarbonyl chloride, 4-bromo
InChI Key ZFFUAQAMUIHRON-UHFFFAOYSA-N
SMILES ClC(=O)C1=CC(Br)=CS1
IUPAC Name 4-bromothiophene-2-carbonyl chloride
Molecular Weight (g/mol) 225.48
PubChem CID 2795483
Formula Weight 225.49
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-thiophenecarbonyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specifica
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.