Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-thiophenecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. B338625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B338625G 25 g
B33865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B338625G Supplier TCI America Supplier No. B338625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16694-18-1
Molecular Formula C5H3BrO2S
Molecular Weight (g/mol) 207.041
MDL Number MFCD03422294
InChI Key HJZFPRVFLBBAMU-UHFFFAOYSA-N
Synonym 4-bromo-2-thiophenecarboxylic acid, 4-bromo-thiophene-2-carboxylic acid, 2-thiophenecarboxylic acid, 4-bromo, 4-bromothiophene-carboxylic acid, 4-bromo-2-thiophene carboxylic acid, 4-bromo-2-carboxythiophene, 4-bromo-2-thiophenecarboxylicacid, 3-bromothiophene-5-carboxylic acid, pubchem5189, 4-bromo-2-thenoic acid
PubChem CID 610409
IUPAC Name 4-bromothiophene-2-carboxylic acid
SMILES C1=C(SC=C1Br)C(=O)O

Specifications

Melting Point 122°C
Color White-Yellow
Quantity 25 g
Formula Weight 207.04
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-thiophenecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.