Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-bromo-3,5-dimethyl-1-phenyl-1h-pyrazole, 97%, Thermo Scientific™

Catalog No. CC48010DA
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC48010DA 1 g
CC48010CB 250 mg
CC48010EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC48010DA Supplier Thermo Scientific Chemicals Supplier No. CC48010DA
Add to Cart

Chemical Identifiers

CAS 51294-75-8
Molecular Formula C11H11BrN2
Molecular Weight (g/mol) 251.127
InChI Key GNXWITGSOFQXDG-UHFFFAOYSA-N
PubChem CID 7039162
IUPAC Name 4-bromo-3,5-dimethyl-1-phenylpyrazole
SMILES CC1=C(C(=NN1C2=CC=CC=C2)C)Br

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 4-bromo-3,5-dimethyl-1-phenyl-1h-pyrazole, 1h-pyrazole,4-bromo-3,5-dimethyl-1-phenyl, 1-phenyl-3,5-dimethyl-4-bromo-1h-pyrazole, 1h-pyrazole, 4-bromo-3,5-dimethyl-1-phenyl
Formula Weight 251.13
Percent Purity 97%
Chemical Name or Material 4-Bromo-3, 5-dimethyl-1-phenyl-1H-pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.