Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-3,5-dimethylisoxazole 98.0+%, TCI America™
SDP

Catalog No. B35195G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 10558-25-5
Molecular Formula C5H6BrNO
Molecular Weight (g/mol) 176.013
MDL Number MFCD00068187
InChI Key GYHZPSUAMYIFQD-UHFFFAOYSA-N
Synonym 4-bromo-3,5-dimethylisoxazole, 3,5-dimethyl-4-bromoisoxazole, isoxazole, 4-bromo-3,5-dimethyl, 4-bromo-dimethylisoxazole, buttpark 33\06-98, 4-bromo-3,5-dimethylisoxazole, tech., pubchem8720, acmc-1bpqs, timtec-bb sbb003784, ksc182i2n
PubChem CID 318421
IUPAC Name 4-bromo-3,5-dimethyl-1,2-oxazole
SMILES CC1=C(C(=NO1)C)Br

Specifications

Color Yellow
Boiling Point 75°C
Quantity 5 g
Formula Weight 176.01
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-3,5-dimethylisoxazole
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.