Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-3-fluoroanisole 98.0+%, TCI America™
SDP

Catalog No. B374325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B374325G 25 g
B37435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B374325G Supplier TCI America Supplier No. B374325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 458-50-4
Molecular Formula C7H6BrFO
Molecular Weight (g/mol) 205.026
MDL Number MFCD01310983
InChI Key XANVIFOBBVAKCY-UHFFFAOYSA-N
Synonym 4-bromo-3-fluoroanisole, 3-fluoro-4-bromo anisole, 3-fluoro-4-bromoanisol, 3-fluoro-4-bromo anisol, 1-bromo-2-fluoro-4-methoxy-benzene, benzene, 1-bromo-2-fluoro-4-methoxy, pharmabridge p-1018, rarechem al mz 0889, zlchem 365, pubchem1970
PubChem CID 3718444
IUPAC Name 1-bromo-2-fluoro-4-methoxybenzene
SMILES COC1=CC(=C(C=C1)Br)F

Specifications

Color Yellow
Boiling Point 216°C
Quantity 25 g
Formula Weight 205.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-3-fluoroanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.