Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Bromo-3'-methylacetanilide 98.0+%, TCI America™
SDP

Catalog No. B314225G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 90914-81-1
Molecular Formula C9H10BrNO
Molecular Weight (g/mol) 228.089
MDL Number MFCD00040822
InChI Key BYZHUFNLXFFINU-UHFFFAOYSA-N
Synonym n-4-bromo-3-methylphenyl acetamide, 4'-bromo-3'-methylacetanilide, 4-bromo-3-methylacetanilide, n-acetyl-4-bromo-m-toluidine, 3-methyl-4-bromoacetanilide, n-acetyl 4-bromo-3-methylaniline, n-4-bromo-3-methylphenyl-acetamide, maybridge1_005107, acmc-209r7e
PubChem CID 738336
IUPAC Name N-(4-bromo-3-methylphenyl)acetamide
SMILES CC1=C(C=CC(=C1)NC(=O)C)Br

Specifications

Melting Point 103°C
Quantity 25 g
Formula Weight 228.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Bromo-3′-methylacetanilide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.