Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-4',4″-dimethyltriphenylamine 95.0+%, TCI America™
SDP

Catalog No. B30891G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30891G 1 g
B30895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30891G Supplier TCI America Supplier No. B30891G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 58047-42-0
Molecular Formula C20H18BrN
Molecular Weight (g/mol) 352.275
InChI Key YMNJJMJHTXGFOR-UHFFFAOYSA-N
Synonym 4-Bromo-N,N-di-p-tolylaniline
PubChem CID 11142701
IUPAC Name N-(4-bromophenyl)-4-methyl-N-(4-methylphenyl)aniline
SMILES CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=CC=C(C=C3)Br

Specifications

Melting Point 102°C
Color White-Yellow
Quantity 1 g
Formula Weight 352.28
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-4′,4′′-dimethyltriphenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.