Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Bromo-4-chlorobutyrophenone, 97%, Thermo Scientific™

Catalog No. AC106851000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC106851000 100 g
AC106850250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC106851000 Supplier Thermo Scientific Chemicals Supplier No. 106851000

Chemical Identifiers

CAS 4559-96-0
Molecular Formula C10H10BrClO
Molecular Weight (g/mol) 261.54
MDL Number MFCD00001006
InChI Key WJKPUMBLABGUCQ-UHFFFAOYSA-N
Synonym 1-4-bromophenyl-4-chlorobutan-1-one, 4'-bromo-4-chlorobutyrophenone, 1-4-bromophenyl-4-chloro-1-butanone, 1-butanone, 1-4-bromophenyl-4-chloro, acmc-20amn1, 4-chloro-p-bromobutyrophenone, 4-chloro-4'-bromobutyrophenone, 4-bromo-4-chlorobutyrophenone, 4/'-bromo-4-chlorobutyrophenone, 4'-bromo-gamma-chlorobutyrophenone
PubChem CID 78317
IUPAC Name 1-(4-bromophenyl)-4-chlorobutan-1-one
SMILES ClCCCC(=O)C1=CC=C(Br)C=C1

Specifications

Melting Point 35°C to 38°C
Flash Point >110°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula BrC6H4CO(CH2)3Cl
Quantity 100 g
Solubility Information Solubility in water: insoluble
Formula Weight 261.54
Percent Purity 97%
Chemical Name or Material 4'-Bromo-4-chlorobutyrophenone
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 224-930-3
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.