Alfa Aesar™ 4'-Bromo-4-chlorobutyrophenone, 98%

Manufacturer: Alfa Aesar™ B2414614

Quantity 25g

 View more versions of this product

Catalog No. AAB2414614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4559-96-0
    Molecular Formula C10H10BrClO
    Formula Weight 261.55
    MDL Number MFCD00001006
    Chemical Name or Material 4'-Bromo-4-chlorobutyrophenone

    Specifications

    Formula Weight 261.55
    Molecular Formula C10H10BrClO
    Melting Point 35° to 38°C
    Quantity 25g
    MDL Number MFCD00001006
    CAS 4559-96-0
    Flash Point >110°C (230°F)
    Chemical Name or Material 4'-Bromo-4-chlorobutyrophenone
    Assay 98%

    Description

    Applications
    4 ft.-Bromo-4-chlorobutyrophenone is used as a pharmaceutical & syntheses material intermediates.

    Solubility
    Insoluble in water.

    Notes
    Storage temperature is 2 - 4°Cand Keep container tightly closed in a dryand well-ventilated place.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit