Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-4'-tert-butylbiphenyl 98.0+%, TCI America™
SDP

Catalog No. B191325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B191325G 25 g
B19135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B191325G Supplier TCI America Supplier No. B191325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 162258-89-1
Molecular Formula C16H17Br
Molecular Weight (g/mol) 289.22
MDL Number MFCD01321141
InChI Key QYNWFBYWVPMMRL-UHFFFAOYSA-N
Synonym 4-bromo-4'-tert-butylbiphenyl, 1,1'-biphenyl,4-bromo-4'-1,1-dimethylethyl, 4-bromo-4'-tert-butyl-1,1'-biphenyl, acmc-209do3, 4-bromo-4'-tertbutylbiphenyl, 4'-bromo-4-tert-butyl-biphenyl, 4-bromo-4/'-tert-butylbiphenyl, 4-bromo-4-tert-butylbiphenyl, 1-4-bromophenyl-4-tert-butylbenzene
PubChem CID 9971389
IUPAC Name 4-bromo-4'-tert-butyl-1,1'-biphenyl
SMILES CC(C)(C)C1=CC=C(C=C1)C1=CC=C(Br)C=C1

Specifications

Melting Point 143°C
Color White-Yellow
Quantity 25 g
Formula Weight 289.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-4′-tert-butylbiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.