Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-alpha-methylbenzylamine, 99%, Thermo Scientific™

Catalog No. ac290990010
Click to view available options
:
1g; Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 24358-62-1
CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C8H10BrN
Refractive Index 1.5665 to 1.5685 (20°C, 589nm)
Linear Formula BrC6H4CH(CH3)NH2
Quantity 1g
MDL Number MFCD00025548
Synonym 1-4-bromophenyl ethylamine, 1-4-bromophenyl ethanamine, 4-bromo-alpha-methylbenzylamine, 4-bromo-alpha-phenethylamine, 1-4-bromophenyl ethan-1-amine, 1-4-bromophenyl ethyl amine, --1-4-bromophenyl ethylamine, +-p-bromo-.alpha.-phenethylamine, 1-amino-1-4-bromophenyl ethane, +/--1-4-bromophenyl ethylamine
InChI Key SOZMSEPDYJGBEK-UHFFFAOYSA-N
SMILES CC(C1=CC=C(C=C1)Br)N
IUPAC Name 1-(4-bromophenyl)ethanamine
Molecular Weight (g/mol) 200.079
PubChem CID 91175
Formula Weight 200.08
Percent Purity ≥98.5%
Chemical Name or Material 4-Bromo-α-methylbenzylamine
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.