Learn More
4'-Bromoacetophenone, 98%
Description
Labeled 4’-Bromoacetophenone possess activity against positive phototaxis of Chlamydomonas cells. A fundamental starting material for organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Labeled 4’-Bromoacetophenone possess activity against positive phototaxis of Chlamydomonas cells. A fundamental starting material for organic synthesis.
Solubility
Soluble in Chloroform. Insoluble in water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent, bases.

Specifications
Specifications
Melting Point | 49°C to 53°C |
Boiling Point | 255°C to 256°C |
Flash Point | >110°C (230°F) |
Quantity | 25 g |
Beilstein | 386015 |
Merck Index | 14,1403 |
Solubility Information | Soluble in Chloroform. Insoluble in water. |
Formula Weight | 199.05 |
Percent Purity | 98% |
Chemical Name or Material | 4'-Bromoacetophenone |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 202-799-3 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.