Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Bromoacetyl)pyridine hydrobromide, 98%

Catalog No. AAH3228506
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 5349-17-7 | C7H7Br2NO | 280.947 g/mol

Design and synthesis of potent, selective, and orally bioavailable tetrasubstituted imidazole inhibitors of p38 mitogen-activated protein kinase 4-(Bromoacetyl)pyridine hydrobromide was used to prepare 2-Phenyl-4-(4-pyridyl)imidazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Design and synthesis of potent, selective, and orally bioavailable tetrasubstituted imidazole inhibitors of p38 mitogen-activated protein kinase 4-(Bromoacetyl)pyridine hydrobromide was used to prepare 2-Phenyl-4-(4-pyridyl)imidazole.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5349-17-7
Molecular Formula C7H7Br2NO
Molecular Weight (g/mol) 280.947
MDL Number MFCD02681893
InChI Key RGALBQILADNMKA-UHFFFAOYSA-N
Synonym 4-bromoacetyl pyridine hydrobromide, 2-bromo-1-pyridin-4-yl ethanone hydrobromide, 2-bromo-1-4-pyridinyl-1-ethanone hydrobromide, 2-bromo-1-pyridin-4-yl ethan-1-one hydrobromide, ethanone, 2-bromo-1-4-pyridinyl-, hydrobromide, 4-2-bromoacetyl pyridine hydrobromide, 4-bromoacetylpyridine hydrobromide, 2-bromo-1-pyrdin-4-ylethan-1-one hydrobromide, 2-bromo-1-4-pyridinyl-1-ethanone hbr, 4-bromoacetyl pyridine hbr
PubChem CID 2776239
IUPAC Name 2-bromo-1-pyridin-4-ylethanone;hydrobromide
SMILES C1=CN=CC=C1C(=O)CBr.Br

Specifications

Melting Point ∽200°C (decomposition)
Odor Weak
Quantity 5 g
UN Number UN3261
Solubility Information Soluble in water.
Formula Weight 280.95
Percent Purity 98%
Chemical Name or Material 4-(Bromoacetyl)pyridine hydrobromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.