Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromoaniline, 96%, pract.

Catalog No. AC402870025
Click to view available options
Quantity:
100 g
500 g
2.5 kg

CAS: 106-40-1 | C6H6BrN | 172.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-40-1
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.02
MDL Number MFCD00007822
InChI Key WDFQBORIUYODSI-UHFFFAOYSA-N
Synonym p-bromoaniline, 4-bromobenzenamine, benzenamine, 4-bromo, p-bromophenylamine, aniline, p-bromo, 4-bromo-aniline, 4-bromobenzeneamine, p-bromo aniline, 4-bromanilinu, 4-bromo-phenylamine
PubChem CID 7807
IUPAC Name 4-bromoaniline
SMILES C1=CC(=CC=C1N)Br

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 60.0°C to 64.0°C
Color Brown to Gray
Density 1.4970g/mL
Boiling Point 230.0°C to 250.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4NH2
Quantity 2.5 kg
Beilstein 12,636
Specific Gravity 1.497
Merck Index 15,1411
Solubility Information Solubility in water: < 1 g / l. Other solubilities: soluble in most organic solvents except cold,petroleum,very soluble in alcohol and ether
Formula Weight 172.02
Percent Purity 96%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Bromoaniline, (pract), 96%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
EINECSNumber 203-393-9
RTECSNumber BW9280000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.