Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromoantipyrine 98.0+%, TCI America™
SDP

Catalog No. B051525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B051525G 25 g
B05155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B051525G Supplier TCI America Supplier No. B051525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5426-65-3
Molecular Formula C11H11BrN2O
Molecular Weight (g/mol) 267.126
MDL Number MFCD00020779
InChI Key QYWWSDVPNCBSIA-UHFFFAOYSA-N
PubChem CID 95032
IUPAC Name 4-bromo-1,5-dimethyl-2-phenylpyrazol-3-one
SMILES CC1=C(C(=O)N(N1C)C2=CC=CC=C2)Br

Specifications

Melting Point 117°C
Color White-Yellow
Quantity 25 g
Formula Weight 267.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromoantipyrine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.