Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromobenzoic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Plastic bottle
586-76-5
C7H5BrO2
201.02
MFCD00002529
TUXYZHVUPGXXQG-UHFFFAOYSA-N
p-bromobenzoic acid,benzoic acid, 4-bromo,p-carboxybromobenzene,benzoic acid, p-bromo,p-bromobenzenecarboxylic acid,4-bromo-benzoic acid,4-brom-benzoesaeure,4-carboxybromobenzene,para bromo benzoic acid,4bromo-benzoic acid
11464
CHEBI:60698
4-bromobenzoic acid
C1=CC(=CC=C1C(=O)O)Br
586-76-5
C7H5BrO2
201.02
MFCD00002529
TUXYZHVUPGXXQG-UHFFFAOYSA-N
p-bromobenzoic acid,benzoic acid, 4-bromo,p-carboxybromobenzene,benzoic acid, p-bromo,p-bromobenzenecarboxylic acid,4-bromo-benzoic acid,4-brom-benzoesaeure,4-carboxybromobenzene,para bromo benzoic acid,4bromo-benzoic acid
11464
CHEBI:60698
4-bromobenzoic acid
C1=CC(=CC=C1C(=O)O)Br
Quantity 100g
Melting Point 251°C to 256°C
Packaging Plastic bottle
Color Brown-Pink to White
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula BrC6H4CO2H
Beilstein 09, 351
Merck Index 15, 1415
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 201.02
Physical Form Crystals or Powder
Percent Purity ≥96.0% (HPLC)
Chemical Name or Material 4-Bromobenzoic acid
Show More Show Less

GHS P Statement Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation. Harmful if swallowed.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.