Learn More
4-Bromobenzonitrile, 98+%
Description
4-Bromobenzonitrile is used as reagents and solvents. Reaction of 2-propanol, 4-bromobenzonitrile, and NaH in the presence of 1.5 mol % Pd2(dba)3 and 3 mol % (S)-(-)2,2?-bis(di-p-tolylphosphino)- 1,1?-binaphthyl at 50°C afforded 4-isopropoxy- benzonitrile in 80% isolated yield. The synthesis of 4-cyanobiphenyl at room temperature in a flow injection microreactor, using a supported catalyst, without the addition of a base gave a product yield of 67±7% (n=6). This was achieved by injecting 4-bromobenzonitrile. The 3-arylation of 2,5-dimethylthiophene with 4-bromopropiophenone, 4-trifluoromethylbromobenzene or 4-bromobenzonitrile gave 3-5 in only 38-50% yields in Palladium-catalysed direct 3-or 4-arylation of thiophene derivatives using aryl bromides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Bromobenzonitrile is used as reagents and solvents. Reaction of 2-propanol, 4-bromobenzonitrile, and NaH in the presence of 1.5 mol % Pd2(dba)3 and 3 mol % (S)-(-)2,2′-bis(di-p-tolylphosphino)- 1,1′-binaphthyl at 50°C afforded 4-isopropoxy- benzonitrile in 80% isolated yield. The synthesis of 4-cyanobiphenyl at room temperature in a flow injection microreactor, using a supported catalyst, without the addition of a base gave a product yield of 67±7% (n=6). This was achieved by injecting 4-bromobenzonitrile. The 3-arylation of 2,5-dimethylthiophene with 4-bromopropiophenone, 4-trifluoromethylbromobenzene or 4-bromobenzonitrile gave 3-5 in only 38-50% yields in Palladium-catalysed direct 3-or 4-arylation of thiophene derivatives using aryl bromides.
Solubility
Very slightly soluble in water(0.2g/L). Soluble in alcohol and ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 111°C to 114°C |
| Density | 1.562 |
| Boiling Point | 235°C to 237°C |
| Flash Point | 130°C (266°F) |
| Quantity | 250 g |
| UN Number | UN3439 |
| Beilstein | 2041518 |
| Solubility Information | Very slightly soluble in water(0.2g/L). Soluble in alcohol and ether. |
| Formula Weight | 182.03 |
| Percent Purity | ≥98% |
| Show More |
Safety and Handling
| Hazard Category | H302+H312-H319 |
| Hazard Statement | GHS H Statement H301-H331-H312-H315-H319-H335 Toxic if swallowed. Toxic if inhaled. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S. |
| EINECSNumber | 210-764-9 |
| RTECSNumber | DI2460000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.