Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromobenzonitrile, 98%

Catalog No. p-73093
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass Bottle
Plastic bottle

CAS: 623-00-7 | C7H4BrN | 182.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-00-7
Molecular Formula C7H4BrN
Molecular Weight (g/mol) 182.02
MDL Number MFCD00001811
InChI Key HQSCPPCMBMFJJN-UHFFFAOYSA-N
Synonym p-bromobenzonitrile, benzonitrile, 4-bromo, benzonitrile, p-bromo, 1-bromo-4-cyanobenzene, 4-bromo benzonitrile, 4-cyano-bromobenzene, 4-bromo-4'-cyanobenzene, 4-cyanobromobenzene, bromobenzonitrile-4, 4-bromocyanobenzene
PubChem CID 12162
IUPAC Name 4-bromobenzonitrile
SMILES BrC1=CC=C(C=C1)C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 111.0°C to 114.0°C
Color White to Yellow
Boiling Point 235.0°C to 237.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula BrC6H4CN
Quantity 10 g
Beilstein 09,354
Solubility Information Solubility in water: 0,2g/L. Other solubilities: soluble in acetone
Formula Weight 182.02
Percent Purity 98%
Physical Form Powder or Flakes
Chemical Name or Material 4-Bromobenzonitrile, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful in contact with skin.
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if y
EINECSNumber 210-764-9
RTECSNumber DI2460000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.