Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromobenzophenone, 98%

Catalog No. aaa1475514
Click to view available options
Quantity:
25 g
100 g
250 g

CAS: 90-90-4 | C13H9BrO | 261.12 g/mol

4-Bromobenzophenone is used in analytical applications such as HPLC and GC. It is also used as a chemical reagent and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzophenone is used in analytical applications such as HPLC and GC. It is also used as a chemical reagent and pharmaceutical intermediate.

Solubility
Insoluble in water but slightly soluble in other solvents such as ethanol, ether and benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-90-4
Molecular Formula C13H9BrO
Molecular Weight (g/mol) 261.12
MDL Number MFCD00000103
InChI Key KEOLYBMGRQYQTN-UHFFFAOYSA-N
Synonym 4-bromobenzophenone, 4-bromophenyl phenyl methanone, p-bromobenzophenone, methanone, 4-bromophenyl phenyl, benzophenone, 4-bromo, p-benzoylbromobenzene, usaf do-3, 4-bromophenyl phenyl ketone, 4-bromophenyl-phenyl-methanone, 4-bromophenyl-phenylmethanone
PubChem CID 7030
SMILES BrC1=CC=C(C=C1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 79°C to 83°C
Boiling Point 350°C
Odor Odorless
Quantity 25 g
Beilstein 1910182
Solubility Information Insoluble in water but slightly soluble in other solvents such as ethanol,ether and benzene.
IUPAC Name (4-bromophenyl)-phenylmethanone
Formula Weight 261.12
Percent Purity 98%
Chemical Name or Material 4-Bromobenzophenone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-024-9
RTECSNumber DJ0350000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.