Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromobenzyl Bromide 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
589-15-1
C7H6Br2
249.93
MFCD00000179
YLRBJYMANQKEAW-UHFFFAOYSA-N
4-bromobenzyl bromide,1-bromo-4-bromomethyl benzene,p-bromobenzyl bromide,alpha,p-dibromotoluene,alpha,4-dibromotoluene,benzene, 1-bromo-4-bromomethyl,4-bromobenzylbromide,unii-3d1ttl8bgn,a,p-dibromotoluene,4-bromo benzyl bromide
68527
1-bromo-4-(bromomethyl)benzene
C1=CC(=CC=C1CBr)Br
589-15-1
C7H6Br2
249.93
MFCD00000179
YLRBJYMANQKEAW-UHFFFAOYSA-N
4-bromobenzyl bromide,1-bromo-4-bromomethyl benzene,p-bromobenzyl bromide,alpha,p-dibromotoluene,alpha,4-dibromotoluene,benzene, 1-bromo-4-bromomethyl,4-bromobenzylbromide,unii-3d1ttl8bgn,a,p-dibromotoluene,4-bromo benzyl bromide
68527
1-bromo-4-(bromomethyl)benzene
C1=CC(=CC=C1CBr)Br
Quantity 100g
Color Tan to White
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 60.0°C to 64.0°C
Boiling Point 115.0°C to 124.0°C (12.0 mmHg)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula BrC6H4CH2Br
Beilstein 05, 308
Merck Index 15, 1416
Solubility Information Solubility in water: may decompose. Other solubilities: soluble in glacial acetic acid,soluble in alcohol,ether,carbon disulfide and,benzene
Formula Weight 249.93
Physical Form Coarse Crystalline Solid or Solid Melt
Percent Purity 98%
Chemical Name or Material 4-Bromobenzyl bromide, 98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-636-5

TSCA : TSCA