Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Bromobenzyl)triphenylphosphonium Bromide 98.0+%, TCI America™
SDP

Catalog No. B524125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B524125G 25 g
B52415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B524125G Supplier TCI America Supplier No. B524125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51044-13-4
Molecular Formula C25H21Br2P
Molecular Weight (g/mol) 512.23
MDL Number MFCD00051922
InChI Key FQJYKXVQABPCRA-UHFFFAOYSA-M
Synonym 4-bromobenzyl triphenylphosphonium bromide, phosphonium, 4-bromophenyl methyl triphenyl-, bromide, 4-bromophenyl methyl triphenylphosphanium bromide, acmc-20apnl, 4-bromobenzyltriphenylphosphoniumbromide, 4-bromobenzyl triphenylphosphoniumbromide, 4-bromobenzyl triphenylphosphonium-bromide, 4-bromobenzyl triphenyl-phosphonium bromide, 4-bromobenzyl triphenyl phosphonium bromide
PubChem CID 2724858
IUPAC Name [(4-bromophenyl)methyl]triphenylphosphanium bromide
SMILES [Br-].BrC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Specifications

Melting Point 275°C
Color White
Quantity 25 g
Formula Weight 512.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (4-Bromobenzyl)triphenylphosphonium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.