Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Bromobutyric acid, 97%
Click to view available options
Quantity:
10 g
50 g
250 g
Description
4-Bromobutyric acid is used as a bifunctional cross-linker and as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromobutyric acid is used as a bifunctional cross-linker and as a pharmaceutical intermediate.
Solubility
Soluble in water and dilute alkali.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at room temperature. It is sensitive to light. Incompatible with strong oxidizing agents
4-Bromobutyric acid is used as a bifunctional cross-linker and as a pharmaceutical intermediate.
Solubility
Soluble in water and dilute alkali.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at room temperature. It is sensitive to light. Incompatible with strong oxidizing agents
Specifications
Specifications
| Melting Point | 31°C to 33°C |
| Boiling Point | 129°C to 131°C (11 mmHg) |
| Flash Point | 115°C (239°F) |
| Odor | Stench |
| Quantity | 10 g |
| UN Number | UN3261 |
| Beilstein | 1743056 |
| Sensitivity | Light sensitive |
| Solubility Information | Soluble in water and dilute alkali. |
| Formula Weight | 167.01 |
| Show More |
Safety and Handling
| Hazard Category | H314 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
| EINECSNumber | 220-083-9 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction