Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromobutyric acid, 97%

Catalog No. AC133471000
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 2623-87-2 | C4H7BrO2 | 167.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2623-87-2
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.00
MDL Number MFCD00002817
InChI Key GRHQDJDRGZFIPO-UHFFFAOYSA-N
Synonym 4-bromobutyric acid, butanoic acid, 4-bromo, 4-bromo-n-butyric acid, butyric acid, 4-bromo, gamma-bromobutyric acid, rarechem al bo 0176, 4-bromobutyricacid, carboxypropyl bromide, 4-bromo-butyric acid, pubchem3754
PubChem CID 75809
IUPAC Name 4-bromobutanoic acid
SMILES OC(=O)CCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 28°C to 32°C
Color Brown
Boiling Point 301°C
Flash Point 124°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula Br(CH2)3COOH
Quantity 100 g
Beilstein 02, 283
Solubility Information Solubility in water: .. Other solubilities: soluble in dilute alkali,soluble in most common organic solvents
Formula Weight 167
Percent Purity 98%
Physical Form Low Melting Crystals
Chemical Name or Material 4-Bromobutyric acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 220-083-9
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.