Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromocinnamic acid, predominantly trans, 97+%

Catalog No. p-7081487
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 1200-07-3 | C9H7BrO2 | 227.06 g/mol

4-Bromocinnamic acid has been used in the preparation of: (E)-β-bromo-4-bromostyrene, 2-amino-7-(piperidin-4-yl)isoquinoline, brominated dansyl derivative (4-bromophenyl)-4-(5-(dimethylamino)naphthalene-1-sulfonamido)butanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromocinnamic acid has been used in the preparation of: (E)-β-bromo-4-bromostyrene, 2-amino-7-(piperidin-4-yl)isoquinoline, brominated dansyl derivative (4-bromophenyl)-4-(5-(dimethylamino)naphthalene-1-sulfonamido)butanoic acid.

Solubility
Soluble in alcohol, ethyl acetate, DMSO. Insoluble in water.

Notes
Store at -20°C. Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1200-07-3
Molecular Formula C9H7BrO2
Molecular Weight (g/mol) 227.06
MDL Number MFCD00004394
InChI Key CPDDDTNAMBSPRN-ZZXKWVIFSA-N
Synonym 4-bromocinnamic acid, p-bromocinnamic acid, 3-4-bromophenyl acrylic acid, bromocinnamic acid,4, 2e-3-4-bromophenyl prop-2-enoic acid, trans-4-bromocinnamic acid, e-3-4-bromophenyl acrylic acid, 2-propenoic acid, 3-4-bromophenyl-, 2e, 4-bromocinnamic acid, predominantly trans, rarechem bk hd c006
PubChem CID 737158
IUPAC Name (E)-3-(4-bromophenyl)prop-2-enoic acid
SMILES OC(=O)\C=C\C1=CC=C(Br)C=C1

Specifications

Melting Point 260°C to 264°C
Odor Odorless
Quantity 5 g
Beilstein 3538823
Solubility Information Soluble in alcohol,ethyl acetate,DMSO. Insoluble in water.
Formula Weight 227.06
Percent Purity ≥98%
Chemical Name or Material 4-Bromocinnamic acid, predominantly trans
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 214-850-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.