Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Bromodiphenyl ether, 99%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
Melting Point | 18.0°C |
Density | 1.4230g/mL |
Boiling Point | 305.0°C |
Flash Point | >113°C |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Refractive Index | 1.6081 |
Linear Formula | BrC6H4OC6H5 |
Quantity | 25 g |
Beilstein | 06,I,105 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Harmful if swallowed. May cause an allergic skin reaction. Causes serious eye damage. Toxic to aquatic life with long lasting effects. |
Precautionary Statement | GHS P Statement Wear protective gloves/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Avoid release to the environment |
EINECSNumber | 202-952-4 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction