Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromomandelic acid, 96%, Thermo Scientific™

Catalog No. AC107110100
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 6940-50-7
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.05
MDL Number MFCD00004232
InChI Key BHZBRPQOYFDTAB-UHFFFAOYNA-N
Synonym 4-bromomandelic acid, 2-4-bromophenyl-2-hydroxyacetic acid, p-bromomandelic acid, p-bromo mandelic acid, 4-bromo-dl-mandelic acid, benzeneacetic acid, 4-bromo-.alpha.-hydroxy, 4-bromophenyl hydroxy acetic acid, 4-bromo-alpha-hydroxyphenylacetic acid, 4-bromo-hydroxybenzeneaceticacid, mandelic acid, p-bromo
PubChem CID 97930
IUPAC Name 2-(4-bromophenyl)-2-hydroxyacetic acid
SMILES OC(C(O)=O)C1=CC=C(Br)C=C1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 115.0°C to 120.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Linear Formula BrC6H4CH(OH)COOH
Quantity 10 g
Beilstein 10, 210
Merck Index 15, 1434
Solubility Information Solubility in water: slightly soluble
Formula Weight 231.05
Percent Purity 96%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 4-Bromomandelic acid, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 230-085-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.