Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Bromomethyl)pyridine hydrobromide, 97%

Catalog No. p-6827172
Click to view available options
Quantity:
5 g
25 g

CAS: 73870-24-3 | C6H7Br2N | 252.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73870-24-3
Molecular Formula C6H7Br2N
Molecular Weight (g/mol) 252.94
MDL Number MFCD01863545
InChI Key VAJUUDUWDNCECT-UHFFFAOYSA-N
Synonym 4-bromomethyl pyridine hydrobromide, 4-bromomethylpyridine hydrobromide, 4-bromomethyl-pyridine hydrobromide, 4-pyridylmethyl bromide hydrobromide, pyridine, 4-bromomethyl-, hydrobromide, 4-bromomethyl pyridine, bromide, 4-bromomethyl-pyridine, 4-bromomethyl pyridinium bromide, acmc-1bbrw
PubChem CID 12752219
IUPAC Name 4-(bromomethyl)pyridine;hydrobromide
SMILES [H+].[Br-].BrCC1=CC=NC=C1

Specifications

Melting Point 189.0°C to 192.0°C
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Quantity 5 g
Formula Weight 252.93
Percent Purity 97%
Chemical Name or Material 4-(Bromomethyl)pyridine hydrobromide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.