Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromomethylbenzenesulfonyl chloride, 95%

Catalog No. AC360540050
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 66176-39-4 | C7H6BrClO2S | 269.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66176-39-4
Molecular Formula C7H6BrClO2S
Molecular Weight (g/mol) 269.55
MDL Number MFCD00156129
InChI Key QXTQWYZHHMQSQH-UHFFFAOYSA-N
Synonym 4-bromomethyl benzenesulfonyl chloride, 4-bromomethyl benzene-1-sulfonyl chloride, 4-bromomethyl benzenesulfonylchloride, alpha-bromo-p-toluenesulphonyl chloride, alpha-bromo-p-toluenesulfonyl chloride, 4-bromomethyl benzenesulphonyl chloride, 4-bromomethyl benzene-1-sulfonylchloride, buttpark 94\04-08, 4-bromomethyl phenyl chlorosulfone, 4-bromomethylene benzenesulfonyl chloride
PubChem CID 2734409
IUPAC Name 4-(bromomethyl)benzenesulfonyl chloride
SMILES C1=CC(=CC=C1CBr)S(=O)(=O)Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 69.0°C to 76.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Argentometry)
Packaging Glass bottle
Linear Formula BrCH2C6H4SO2Cl
Quantity 5 g
Formula Weight 269.55
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromomethylbenzenesulfonyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.