Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromophenol, 99%

Catalog No. aaa1487322
Click to view available options
Quantity:
100 g
500 g
2500 g

CAS: 106-41-2 | C6H5BrO | 173.009 g/mol

4-Bromophenol is used as an intermediate in medicine grade. It is also used as a phenol that is used in Stille reactions. It is used in the intermediate of organic synthesis, flame retardant and pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromophenol is used as an intermediate in medicine grade. It is also used as a phenol that is used in Stille reactions. It is used in the intermediate of organic synthesis, flame retardant and pharmaceutical industry.

Solubility
It is insoluble in water. It is soluble in 5%ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Store under inert gas. Air sensitive. Keep away from Oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-41-2
Molecular Formula C6H5BrO
Molecular Weight (g/mol) 173.009
MDL Number MFCD00002313
InChI Key GZFGOTFRPZRKDS-UHFFFAOYSA-N
Synonym p-bromophenol, phenol, 4-bromo, p-bromohydroxybenzene, 4-bromo-phenol, p-bromophenic acid, phenol, p-bromo, para-bromophenol, 4-bromo phenol, unii-lao4j0183i, ccris 632
PubChem CID 7808
IUPAC Name 4-bromophenol
SMILES C1=CC(=CC=C1O)Br

Specifications

Melting Point 62°C to 66°C
Density 1.840
Boiling Point 235°C to 238°C
Flash Point >110°C (230°F)
Assay Percent Range 99%
Quantity 100 g
UN Number UN2811
Beilstein 1680024
Sensitivity Light sensitive
Merck Index 14,1428
Solubility Information It is insoluble in water. It is soluble in 5%ethanol.
Formula Weight 173.01
Percent Purity 99%
Chemical Name or Material 4-Bromophenol
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H312-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 203-394-4
RTECSNumber SJ7960000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.