Alfa Aesar™ 4-Bromosalicylaldehyde, 97%

Manufacturer: Alfa Aesar™ H5485003

Quantity 1g

 View more versions of this product

Catalog No. AAH5485003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 22532-62-3
    Molecular Formula C7H5BrO2
    Formula Weight 201.02
    MDL Number MFCD06252606
    UN Number UN3077
    Synonym 4-Bromo-2-hydroxybenzaldehyde
    Chemical Name or Material 4-Bromosalicylaldehyde

    Specifications

    Formula Weight 201.02
    Melting Point 50° to 54°C
    Quantity 1g
    Flash Point >110°C (230°F)
    Molecular Formula C7H5BrO2
    MDL Number MFCD06252606
    UN Number UN3077
    Synonym 4-Bromo-2-hydroxybenzaldehyde
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H318
    Toxic if swallowed.
    Causes serious eye damage.
    CAS 22532-62-3
    Health Hazard 3 GHS P Statement
    P280-P301+P310-P305+P351+P338-P310-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Chemical Name or Material 4-Bromosalicylaldehyde
    Assay 97%

    Description

    Applications
    It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

    Solubility
    Soluble in methanol.

    Notes
    Strong bases and oxidizing agents are to be avoided.

    GHS H Statement
    H301-H318
    Toxic if swallowed.
    Causes serious eye damage.

    GHS P Statement
    P280-P301+P310-P305+P351+P338-P310-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit