Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromostyrene, 96%, stabilized

Catalog No. p-4510264
Click to view available options
Quantity:
10 mL
25 mL
100 mL
Packaging:
Glass bottle

CAS: 2039-82-9 | C8H7Br | 183.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2039-82-9
Molecular Formula C8H7Br
Molecular Weight (g/mol) 183.05
MDL Number MFCD00000110
InChI Key WGGLDBIZIQMEGH-UHFFFAOYSA-N
Synonym 4-bromostyrene, 1-bromo-4-vinylbenzene, p-bromostyrene, benzene, 1-bromo-4-ethenyl, styrene, p-bromo, 1-4-bromophenyl ethylene, unii-tvk4s80xlu, p-bromo styrene, 1-bromo-4-vinyl-benzene, tvk4s80xlu
PubChem CID 16263
IUPAC Name 1-bromo-4-ethenylbenzene
SMILES BrC1=CC=C(C=C)C=C1

Specifications

Melting Point 4.5°C
Color Colorless to Yellow
Density 1.4000g/mL
Boiling Point 88.0°C to 89.0°C (16.0 mmHg)
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5930 to 1.5950
Linear Formula H2C=CHC6H4Br
Quantity 10 mL
Beilstein 05,II,367
Specific Gravity 1.4
Stabilizer 100 to 500ppm 4-tert.-butylcatechol
Formula Weight 183.05
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 4-Bromostyrene
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 218-022-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.