Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromostyrene (stabilized with TBC) 95.0+%, TCI America™
SDP

Catalog No. p-7119397
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 2039-82-9
Molecular Formula C8H7Br
Molecular Weight (g/mol) 183.05
MDL Number MFCD00000110
InChI Key WGGLDBIZIQMEGH-UHFFFAOYSA-N
Synonym 4-bromostyrene, 1-bromo-4-vinylbenzene, p-bromostyrene, benzene, 1-bromo-4-ethenyl, styrene, p-bromo, 1-4-bromophenyl ethylene, unii-tvk4s80xlu, p-bromo styrene, 1-bromo-4-vinyl-benzene, tvk4s80xlu
PubChem CID 16263
IUPAC Name 1-bromo-4-ethenylbenzene
SMILES BrC1=CC=C(C=C)C=C1

Specifications

Color Yellow
Boiling Point 81°C
Quantity 5 g
Formula Weight 183.05
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromostyrene (stabilized with TBC)
TSCA No
Recommended Storage Freezer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.