Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromothioanisole 99.0+%, TCI America™
SDP

Catalog No. B2024250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2024250G 250 g
B202425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2024250G Supplier TCI America Supplier No. B2024250G

Chemical Identifiers

CAS 104-95-0
Molecular Formula C7H7BrS
Molecular Weight (g/mol) 203.10
MDL Number MFCD00000102
InChI Key YEUYZNNBXLMFCW-UHFFFAOYSA-N
Synonym 4-bromothioanisole, 1-bromo-4-methylthio benzene, p-bromophenyl methyl sulfide, 4-bromophenyl methyl sulfide, benzene, 1-bromo-4-methylthio, 1-bromo-4-methylsulfanyl benzene, p-bromothioanisole, 4-bromo-thioanisole, methyl 4-bromophenylsulfide, p-bromo thioanisole
PubChem CID 66037
IUPAC Name 1-bromo-4-(methylsulfanyl)benzene
SMILES CSC1=CC=C(Br)C=C1

Specifications

Melting Point 38°C
Color White-Yellow
Boiling Point 250°C
Quantity 250 g
Formula Weight 203.10
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromothioanisole
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.