Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromothiophene-3-carboxaldehyde 97.0+%, TCI America™
SDP

Catalog No. B4126200MG
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 18791-78-1
Molecular Formula C5H3BrOS
Molecular Weight (g/mol) 191.04
MDL Number MFCD01318960
InChI Key ZDVHJQOCVBTMOF-UHFFFAOYSA-N
Synonym 4-Bromo-3-formylthiophene
PubChem CID 2764207
IUPAC Name 4-bromothiophene-3-carbaldehyde
SMILES BrC1=CSC=C1C=O

Specifications

Color Yellow
Boiling Point 120°C
Quantity 200 mg
Formula Weight 191.04
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromothiophene-3-carboxaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.