Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromothiophenol, 98%

Catalog No. p-7080835
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 106-53-6 | C6H5BrS | 189.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-53-6
Molecular Formula C6H5BrS
Molecular Weight (g/mol) 189.07
MDL Number MFCD00004845
InChI Key FTBCOQFMQSTCQQ-UHFFFAOYSA-N
Synonym 4-bromothiophenol, benzenethiol, 4-bromo, p-bromothiophenol, p-bromobenzenethiol, 4-bromobenzene-1-thiol, 4-bromo-benzenethiol, 4-bromo thiophenol, p-bromophenyl thiol, benzenethiol, p-bromo, unii-p3wuo03x17
PubChem CID 66049
IUPAC Name 4-bromobenzenethiol
SMILES C1=CC(=CC=C1S)Br

Specifications

Melting Point 72°C to 78°C
Boiling Point 239°C
Odor Unpleasant
Quantity 10 g
UN Number UN2923
Beilstein 507031
Sensitivity Air Sensitive
Formula Weight 189.08
Percent Purity 98%
Chemical Name or Material 4-Bromothiophenol
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 203-407-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.