Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromotoluene, 99%

Catalog No. p-77379
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle

CAS: 106-38-7 | C7H7Br | 171.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-38-7
Molecular Formula C7H7Br
Molecular Weight (g/mol) 171.04
MDL Number MFCD00000109
InChI Key ZBTMRBYMKUEVEU-UHFFFAOYSA-N
Synonym 4-bromotoluene, p-bromotoluene, 4-methylbromobenzene, p-tolyl bromide, benzene, 1-bromo-4-methyl, toluene, p-bromo, p-methylphenyl bromide, p-methylbromobenzene, 1-methyl-4-bromobenzene, 4-bromo-1-methylbenzene
PubChem CID 7805
IUPAC Name 1-bromo-4-methylbenzene
SMILES CC1=CC=C(Br)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 26°C to 29°C
Color White
Density 1.3900g/mL
Boiling Point 184°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.549
Linear Formula CH3C6H4Br
Quantity 100 mL
Beilstein 05, 305
Specific Gravity 1.39
Merck Index 14, 1439
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohol,ether and benzene
Formula Weight 171.04
Percent Purity 99%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 4-Bromotoluene, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-391-8
RTECSNumber XS7965600
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.