Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Butoxy-4'-cyanobiphenyl 98.0+%, TCI America™
SDP

Catalog No. B430025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B430025G 25 g
B43005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B430025G Supplier TCI America Supplier No. B430025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 52709-87-2
Molecular Formula C17H17NO
Molecular Weight (g/mol) 251.33
MDL Number MFCD00183755
InChI Key KPQVQWUNELODQE-UHFFFAOYSA-N
PubChem CID 104292
IUPAC Name 4'-butoxy-[1,1'-biphenyl]-4-carbonitrile
SMILES CCCCOC1=CC=C(C=C1)C1=CC=C(C=C1)C#N

Specifications

Melting Point 77°C
Color White
Quantity 25 g
Synonym 4′-Butyloxy-4-biphenylcarbonitrile, 4OCB
Formula Weight 251.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Butoxy-4′-cyanobiphenyl
RTECSNumber DV1764000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.