Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Butoxyaniline 98.0+%, TCI America™
SDP

Catalog No. B100325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B100325G 25 g
B10035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B100325G Supplier TCI America Supplier No. B100325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4344-55-2
Molecular Formula C10H15NO
Molecular Weight (g/mol) 165.236
MDL Number MFCD00007866
InChI Key UBRIHZOFEJHMIT-UHFFFAOYSA-N
Synonym p-butoxyaniline, benzenamine, 4-butoxy, p-butyloxyaniline, 4-n-butoxyaniline, aniline, p-butoxy, p-aminophenol n-butyl ether, 4-butyloxybenzenamine, aminophenyl-n-butyl ether, 4-butyloxyaniline, 4-butyloxy aniline
PubChem CID 20352
IUPAC Name 4-butoxyaniline
SMILES CCCCOC1=CC=C(C=C1)N

Specifications

Color Brown
Boiling Point 157°C
Quantity 25 g
Formula Weight 165.24
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Butoxyaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.