Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Butyl-4'-cyanobiphenyl 98.0+%, TCI America™
SDP

Catalog No. b49235g Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g

Catalog No. B49235G

Supplier: TCI America B49235G

Only null left
Add to Cart
Add to Cart

Specifications

CAS 52709-83-8
Melting Point 49°C
Color White
Boiling Point 195°C
Molecular Formula C17H17N
Quantity 5 g
MDL Number MFCD00506009
Synonym 4'-butyl-1,1'-biphenyl-4-carbonitrile, 4-butyl-4'-cyanobiphenyl, 4'-butyl-4-biphenylcarbonitrile, 4-cyano-4'-butylbiphenyl, 1,1'-biphenyl-4-carbonitrile, 4'-butyl, 4-n-butyl-4'-cyanobiphenyl, 4-4-butylphenyl benzonitrile, 4'-butyl 1,1'-biphenyl-4-carbonitrile, 4'-butyl-4-cyanobiphenyl
InChI Key PJPLBHHDTUICNN-UHFFFAOYSA-N
SMILES CCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C#N
IUPAC Name 4-(4-butylphenyl)benzonitrile
Molecular Weight (g/mol) 235.33
PubChem CID 104288
Formula Weight 235.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Butyl-4′-cyanobiphenyl
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.