Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Butylacetophenone 97.0+%, TCI America™
SDP

Catalog No. B1000500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1000500G 500 g
B100025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1000500G Supplier TCI America Supplier No. B1000500G

Chemical Identifiers

CAS 37920-25-5
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.26
MDL Number MFCD00017500
InChI Key MQESVSITPLILCO-UHFFFAOYSA-N
Synonym 4'-butylacetophenone, 1-4-butylphenyl ethanone, ethanone, 1-4-butylphenyl, p-butylacetophenone, 1-4-butylphenyl ethan-1-one, p-n-butylacetophenone, 4'-n-butylacetophenone, unii-6738cf9z8v, 1-acetyl-4-butylbenzene, 1-4-butyl-phenyl ethanone
PubChem CID 123470
IUPAC Name 1-(4-butylphenyl)ethan-1-one
SMILES CCCCC1=CC=C(C=C1)C(C)=O

Specifications

Color Yellow
Boiling Point 270°C
Quantity 500 g
Formula Weight 176.26
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-Butylacetophenone
EINECSNumber (3)-3540
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.