Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Butylcyclohexanecarboxylic Acid (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. B298625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B298625G 25 g
B29865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B298625G Supplier TCI America Supplier No. B298625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 71101-89-8
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD00460757
InChI Key BALGERHMIXFENA-UHFFFAOYSA-N
PubChem CID 2060518
IUPAC Name 4-butylcyclohexane-1-carboxylic acid
SMILES CCCCC1CCC(CC1)C(=O)O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 184.28
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Butylcyclohexanecarboxylic Acid (cis- and trans- mixture)
EINECSNumber (3)-3723
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.